Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14912

Sigma-Aldrich

tert-Dodecanethiol (mixture of isomers)

for synthesis

Synonym(s):

tert-Dodecanethiol (mixture of isomers), Lauryl mercaptan, Dodecyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H26S
CAS Number:
Molecular Weight:
202.40
UNSPSC Code:
12352105
EC Index Number:
246-619-1

vapor pressure

1.33 hPa ( 25.5 °C)

form

liquid

autoignition temp.

350 °C

potency

6800 mg/kg LD50, oral (Rat)
12600 mg/kg LD50, skin (Rabbit)

bp

233 °C/1013 hPa

mp

-45 °C

transition temp

flash point 98 °C

solubility

<0.1 g/L

density

0.856 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C12H26S/c1-9(2)10(3,4)11(5,6)12(7,8)13/h9,13H,1-8H3

InChI key

YAJYJWXEWKRTPO-UHFFFAOYSA-N

Analysis Note

Assay (of S): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 0.853 - 0.858
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service