Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14906

Sigma-Aldrich

3-Allyloxy-2-hydroxypropane sulfonate sodium salt

for synthesis

Synonym(s):

3-Allyloxy-2-hydroxypropane sulfonate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NaO5S
CAS Number:
Molecular Weight:
218.20
MDL number:
UNSPSC Code:
12352107
EC Index Number:
258-004-5

vapor pressure

23 hPa ( 20 °C)

Quality Level

form

liquid

potency

18800 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

pH

6-9 (20 °C, 111 g/L in H2O)

mp

9 °C

transition temp

flash point >100 °C

density

1.17 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H12O5S.Na/c1-2-3-11-4-6(7)5-12(8,9)10;/h2,6-7H,1,3-5H2,(H,8,9,10);/q;+1/p-1

InChI key

CJGJYOBXQLCLRG-UHFFFAOYSA-M

Analysis Note

Assay (ionoc exchange): 38.0 - 42.0 %

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service