Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.14082

Sigma-Aldrich

(S)-(+)-2-Amino-3-methyl-1-butanol

for synthesis

Synonym(s):

(S)-(+)-2-Amino-3-methyl-1-butanol, L-Valinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH3)CHCH(NH2)CH2OH
CAS Number:
Molecular Weight:
103.16
MDL number:
UNSPSC Code:
12352104
EC Index Number:
217-975-5

Quality Level

Assay

≥97.5% (GC)

form

crystals

bp

91-92 °C/13 hPa

mp

31-32 °C

transition temp

flash point 78 °C

density

0.93 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C5H13NO/c1-4(2)5(6)3-7/h4-5,7H,3,6H2,1-2H3

InChI key

NWYYWIJOWOLJNR-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.932 - 0.934
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service