Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.11720

Sigma-Aldrich

Kryptofix® 21

for synthesis

Synonym(s):

Kryptofix® 21, 1,4,10-Trioxa-7,13-diazacyclopentadecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22N2O3
CAS Number:
Molecular Weight:
218.29
MDL number:
UNSPSC Code:
12352005
EC Index Number:
250-530-3

form

powder

mp

89-92 °C

storage temp.

2-30°C

InChI

1S/C10H22N2O3/c1-5-13-6-2-12-4-8-15-10-9-14-7-3-11-1/h11-12H,1-10H2

InChI key

STHIZMRUXPMSCW-UHFFFAOYSA-N

Analysis Note

Assay (perchloric acid titration): ≥ 97
Identity (IR-spectrum): passes test

Legal Information

Kryptofix is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service