Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.08540

Sigma-Aldrich

Ethyl pentanoate

for synthesis

Synonym(s):

Ethyl pentanoate, Ethyl valerate, Valeric acid ethyl ester, Pentanoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3COOC2H5
CAS Number:
Molecular Weight:
130.18
MDL number:
UNSPSC Code:
12352108
EC Index Number:
208-726-1

vapor pressure

3 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

400 °C

expl. lim.

1 % (v/v)

refractive index

n20/589 1.399

bp

145 °C/1013 hPa

mp

-91 °C

transition temp

flash point 35 °C

solubility

2.23 g/L

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H14O2/c1-3-5-6-7(8)9-4-2/h3-6H2,1-2H3

InChI key

ICMAFTSLXCXHRK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.874 - 0.875
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service