Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.02400

Sigma-Aldrich

3-Bromotoluene

for synthesis

Synonym(s):

3-Bromotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3-(Br)C6H4CH3
CAS Number:
Molecular Weight:
171.03
MDL number:
UNSPSC Code:
12352101
EC Index Number:
209-702-3

vapor pressure

2.04 hPa ( 25 °C)

Assay

≥98.0% (GC)

form

liquid

potency

1540 mg/kg LD50, oral (Rat)

mp

-40 °C

transition temp

flash point 60 °C

density

1.405 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C7H7Br/c1-6-3-2-4-7(8)5-6/h2-5H,1H3

InChI key

WJIFKOVZNJTSGO-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.405 - 1.415
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(C)

60 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service