Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00476

Sigma-Aldrich

4-Chlorobenzoic acid

for synthesis

Synonym(s):

4-Chlorobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(Cl)C6H4COOH
CAS Number:
Molecular Weight:
156.57
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-805-9

Assay

≥98.0% (acidimetric)

form

powder

autoignition temp.

545 °C

potency

1170 mg/kg LD50, oral (Rat)

pH

4.3 (20 °C, 0.08 g/L in H2O)

bp

274-276 °C/1013 hPa

mp

237-240 °C

transition temp

flash point 238 °C

solubility

0.08 g/L

density

1.54 g/cm3 at 24 °C

bulk density

350 kg/m3

storage temp.

2-30°C

InChI

1S/C7H5ClO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)

InChI key

XRHGYUZYPHTUJZ-UHFFFAOYSA-N

Application

4-Chlorobenzoic acid for synthesis. CAS 74-11-3, chemical formula 4-(Cl)C₆H₄COOH.

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 237 °C
Melting range (upper value): ≤ 241 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(C)

238 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service