Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

6.17594

Supelco

Barium nitrate

purified grade

Synonym(s):

Barium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(NO3)2
CAS Number:
Molecular Weight:
261.34
MDL number:
UNSPSC Code:
12352302

grade

purified grade

InChI

1S/Ba.2NO3/c;2*2-1(3)4/q+2;2*-1

InChI key

IWOUKMZUPDVPGQ-UHFFFAOYSA-N

Pictograms

Flame over circleSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service