Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

6.09984

Supelco

Sulfuric acid

c(H2SO4) = 0.05 mol/l (0.1N), concentrate for preparing 0.5 L volumetric solution for titration, Titrisol®

Synonym(s):

Sulfuric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352322

product name

Sulfuric acid, for 500 ml, c(H2SO4) = 0.05 mol/l (0.1N) Titrisol

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application



  • Isolation, development and validation of HPTLC method for the estimation of β-carotene from Gymnosporia senegalensis (Lam.) Loes.: This study developed and validated a High-Performance Thin Layer Chromatography (HPTLC) method to estimate β-carotene, using n-Hexane as a solvent, enhancing the reliability and precision of β-carotene quantification in plant extracts (Jain et al., 2023).


  • Antimicrobial, antioxidant, and sun protection potential of the isolated compounds from Spermacoce princeae (K. Schum).: This research explored the bioactive compounds of Spermacoce princeae using n-Hexane extraction, highlighting its effectiveness in isolating potent antimicrobial and antioxidant agents (Sekandi et al., 2023).


  • Stable isotope characterization of tobacco products: A determination of synthetic or natural nicotine authenticity.: The study employed n-Hexane for stable isotope analysis, providing critical insights into the authenticity of nicotine in tobacco products through advanced mass spectrometry techniques (Han et al., 2023).


  • Kinetic studies on the extraction of oil from a new feedstock (Chukrasia tabularis L. seed) for biodiesel production using a heterogeneous catalyst.: This research focused on the kinetics of oil extraction using n-Hexane, optimizing the process for biodiesel production and offering a sustainable alternative feedstock (Eswaramoorthi et al., 2023).


  • Photocatalytic oxidation of selected gas-phase VOCs using UV light, TiO2, and TiO2/Pd.: The study utilized n-Hexane to investigate the photocatalytic degradation of volatile organic compounds (VOCs), presenting a viable method for air purification and environmental remediation (Fujimoto et al., 2017).




Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service