Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

559300

Sigma-Aldrich

Safingol

A cell-permeable and reversible lyso-sphingolipid protein kinase C (PKC) inhibitor that competitively interacts at the regulatory phorbol binding domain of PKC.

Synonym(s):

Safingol, L- threo-Dihydrosphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H39NO2
CAS Number:
Molecular Weight:
301.51
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

ethanol: 10 mg/mL
DMSO: 5 mg/mL
methanol: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C18H39NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)17(19)16-20/h17-18,20-21H,2-16,19H2,1H3

InChI key

OTKJDMGTUTTYMP-UHFFFAOYSA-N

General description

A cell-permeable and reversible lyso-sphingolipid protein kinase C (PKC) inhibitor that competitively interacts at the regulatory phorbol binding domain of PKC. Inhibits enzymatic activity and 3H-phorbol dibutyrate binding of purified rat brain PKC (IC50 = 37.5 µM and 31 µM, respectively). Inhibits human PKCα, the major overexpressed isoenzyme in MCF-7 DOXR cells (IC50 = 40 µM). Enhances the cytotoxic effect of the chemotherapeutic agent Mitomycin C (MMC; Cat. Nos. 47589 and 475820) in gastric cancer cells by promoting drug-induced apoptosis.
A cell-permeable and reversible lyso-sphingolipid protein kinase C (PKC) inhibitor that competitively interacts at the regulatory phorbol binding domain of PKC. Inhibits enzymatic activity and 3H-phorbol dibutyrate binding of purified rat brain PKC (IC50 = 37.5 µM and 31 µM, respectively). Inhibits human PKCα, the major overexpressed isoenzyme in MCF-7 DOXR cells (IC50 = 40 µM). Enhances the cytotoxic effect of the chemotherapeutic agent mitomycin C (MMC; Cat. No. 47589) in gastric cancer cells by promoting drug-induced apoptosis.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Enzymatic activity and 3H-phorbol dibutyrate binding of purified rat brain PKC
Product does not compete with ATP.
Reversible: yes
Target IC50: 37.5 µM and 31 µM, inhibiting enzymatic activity and 3H-phorbol dibutyrate binding of purified rat brain PKC, respectively

Warning

Toxicity: Irritant (B)

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kedderis, L.B., et al. 1995. Fundam. Appl. Toxicol.25, 201.
Sachs, C.W., et al. 1995. J. Biol. Chem. 270, 26639.
Schwartz, G.K., et al. 1995. J. Natl. Cancer Inst.87,1394.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service