Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

539695

Sigma-Aldrich

PME-1 Inhibitor, AMZ-30

The PME-1 Inhibitor, AMZ-30 controls the biological activity of PME-1. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

PME-1 Inhibitor, AMZ-30, ( E)-2-(4-Fluorophenylsulfonyl)-3-(1-(3-nitrophenylsulfonyl)-1H-pyrrol-2-yl)-acrylonitrile, NIH Probe ML136

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H13FN3O6S2
Molecular Weight:
462.45
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

2-8°C

General description

A cell-permeable sulfonyl acrylonitrile compound that acts as a potentand irreversible inhibitor of protein phosphatase methylesterase-1(PME-1; IC50 = 600 nM and 2.5 µM in human cell lysates and in HEK 293T cells, respectively) with 100-fold greater selectivity over other serine hydrolases; does not function as a general thiol-reactive probe. Shown to lower demethylated PP2A levels by ~80% at 20 µM with elevated methylated PP2A (protein phosphatase 2A) form in PME-1 overexpressing HEK 293T cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service