Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

539530

Sigma-Aldrich

Anti-PP2Bα (482-494) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Calcineurin, Anti-PP2Bα, Anti-PP 2Bα

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... PPP3CA(5530)

General description

Recognizes the ~60 kDa PP2B catalytic subunit.
  • Antibody Target Gene Symbol: PPP3CA
  • Target Synonym: 2900074D19Rik, AI841391, AW413465, Calcineurin, Calcineurin A Alpha, CALN, CALNA, CALNA1, CCN1, CN, CnA, CnA-alpha, CNA1, MGC106804, Pp2b Subunit A, PP2B-A ALPHA, PPP2B, PPP3CA alpha isoform
  • Entrez Gene Name: protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform
  • Human Entrez ID: 5530
  • Mu Entrez ID: 19055
  • Rat Entrez ID: 24674
  • Immunogen

    Mammalian
    a synthetic peptide (RMPPRRDAMPSDA) corresponding to amino acids 482-494 of the mammalian PP2B catalytic subunit, conjugated to KLH

    Application

    Immunoblotting (1:500)

    Immunocytochemistry (1:500)

    Warning

    Toxicity: Standard Handling (A)

    Physical form

    In 10 mM Tris-HCl, 3 mM glycine, pH 7.5.

    Reconstitution

    Following initial thaw, aliquot and freeze (-70°C).

    Other Notes

    No cross-reactivity to other protein phosphatases is detected. Variables associated with assay conditions will dictate the proper working dilution.

    Legal Information

    CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

    Not finding the right product?  

    Try our Product Selector Tool.

    Storage Class Code

    10 - Combustible liquids

    WGK

    WGK 1

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service