Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

537076

Sigma-Aldrich

Proprotein Convertase Inhibitor

The Proprotein Convertase Inhibitor controls the biological activity of Proprotein Convertase. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

Proprotein Convertase Inhibitor, PACE4 Inhibitor, Furin-like Proprotein Convertase Inhibitor, Furin Inhibitor III, PC1/3 Inhibitor, PC4 Inhibitor, PC5/6 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H55N15O4
Molecular Weight:
749.91
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

potency

16 pM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable tripeptide derivative containing an Arg-X-Arg motive that acts as a highly potent and reversible furin inhibitor (Ki = 16 pM). Also shown to inhibit several other pro-protein convertases, such as PC1/3, PC4, PAEC4, PC5/6 with almost similar potency, but does not affect PC2, PC7, and trypsin-like serine proteases in any significant manner. Acts as a very weak inhibitor of plasmin (Ki = 7.3 µM). Exhibits greater stability in cells. Shown to block hemagglutinin cleavage (40% at 1 µM) and fowl plague virus (FPV) viral propagation in a dose-dependent manner. Exhibits minimal toxicity even at high concentrations (25-50 µM) and protects human epidermoid carcinoma (HEp-2) cells from toxic effects of Shiga toxin.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Furin, PAEC4, PC1/3, PC4, and PC5/6
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Becker, G., et al. 2012. J Biol Chem287, 21992.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi He et al.
Osteoarthritis and cartilage open, 3(2), 100162-100162 (2021-04-14)
Cartilage degradation is a hallmark of osteoarthritis (OA). Aggrecan, a major proteoglycan of articular cartilage extracellular matrix (ECM), is degraded by ADAMTS-5 resulting in the release of ARGS-G2 fragments to synovial fluid and circulation. The aim was to quantify ARGS-G2
Belinda Yau et al.
Scientific reports, 9(1), 19466-19466 (2019-12-21)
A threonine-to-Isoleucine (Thr52Ile) mutation within the pro-domain of the Sorcs1 gene was positionally cloned as the gene underlying a quantitative trait locus that affects fasting insulin levels in mice. In humans, genome-wide association studies and linkage studies have shown that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service