Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.30648

Sigma-Aldrich

SIRT2 Inhibitor, AEM2

Synonym(s):

SIRT2 Inhibitor, AEM2, Sirtuin 2 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27N3O
CAS Number:
Molecular Weight:
409.52
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

3.8 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell permeable methanophenanthridine derivative that acts a selective inhibitor of SIRT2 (IC50 = 3.8 µM) and exhibits ~30 fold greater selectivity over SIRT1 (IC50 >100 µM) and SIRT3. Blocks SIRT2-induced p53 deacetylation and thereby elevates p53 levels and causes increased expression of CDKN1A, PUMA, and NOXA in non-small cell lung cancer (NSCLC) A549 cells. Also shown to reduce NSCLC cell viability by 50% (~20 µM) and inhibit their proliferation. Sensitizes A549 cancer cell line to etoposide (~1 µM) induced DNA damage in a p53-dependent manner.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
SIRT2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 1 month at -70°C.
Use only fresh DMSO for reconstitution.

Other Notes

Hoffmann G., et al. 2014. J. Biol. Chem.289, 5208.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gesine Hoffmann et al.
The Journal of biological chemistry, 289(8), 5208-5216 (2014-01-01)
Sirtuin 2 (SIRT2) is an NAD(+)-dependent protein deacetylase whose targets include histone H4 lysine 16, p53, and α-tubulin. Because deacetylation of p53 regulates its effect on apoptosis, pharmacological inhibition of SIRT2-dependent p53 deacetylation is of great therapeutic interest for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service