Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.08739

Millipore

Cycloheximide

InSolution, ≥98%

Synonym(s):

InSolution Cycloheximide, High Purity, 4-(( R)-2-((1 S,3 S,5 S)-3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl)piperidine-2,6-dione, Actidione, 4-((R)-2-((1S,3S,5S)-3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl)piperidine-2,6-dione, Actidione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NO4
CAS Number:
Molecular Weight:
281.35
MDL number:
UNSPSC Code:
12352005

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−20°C

InChI

1S/C15H23NO4/c1-8-3-9(2)15(20)11(4-8)12(17)5-10-6-13(18)16-14(19)7-10/h8-12,17H,3-7H2,1-2H3,(H,16,18,19)/t8-,9-,11-,12+/m0/s1

InChI key

YPHMISFOHDHNIV-FSZOTQKASA-N

Biochem/physiol Actions

Cell permeable: yes
Primary Target
translocase enzyme
Reversible: yes

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Physical form

A 300 mM (100 mg/1.185 ml) sterile-filtered solution of Cycloheximide, High Purity (Cat. No. 239764) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C.

Other Notes

Christner, C., et al. 1999. J. Med. Chem.42, 3615.
Lu, Q., et al. 1996. Arch. Biochem. Biophys.334, 175.

Chow, S.C., et al. 1995. Exp. Cell Res.216, 149.
Cotter, T.G., et al. 1992. Anticancer Res.12, 773.
Takano, Y.S., et al. 1991. J. Pathol.163, 329.
Waring, P. 1990. J. Biol. Chem.265, 14476.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Muta. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service