Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420210

Sigma-Aldrich

Jervine

A cell-permeable steroidal alkaloid similar to cyclopamine that displays teratogenic effects and induces cyclopia by blocking Sonic Hedgehog signaling.

Synonym(s):

Jervine, (3β, 23β)-17,23-Epoxy-3-hydroxyveratraman-11-one, 11-Ketocyclopamine, Shh Signaling Antagonist III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H39NO3
CAS Number:
Molecular Weight:
425.60
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

ethanol: 10 mg/mL
DMSO: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C27H39NO3/c1-14-11-21-24(28-13-14)16(3)27(31-21)10-8-19-20-6-5-17-12-18(29)7-9-26(17,4)23(20)25(30)22(19)15(27)2/h5,14,16,18-21,23-24,28-29H,6-13H2,1-4H3

InChI key

CLEXYFLHGFJONT-UHFFFAOYSA-N

General description

A cell-permeable steroidal alkaloid similar to cyclopamine (Cat. No. 239803) that displays teratogenic effects and induces cyclopia by blocking shh (Sonic Hedgehog) signaling (IC50 = ~ 500-700 nM in s12 cells). Tomatidine (Cat. No. 614350) serves as a suitable negative control.
A cell-permeable steroidal alkaloid similar to cyclopamine (Cat. No. 239803) that displays teratogenic effects and induces cyclopia by blocking shh (Sonic Hedgehog) signaling (IC50 ~500-700 nM in s12 cells). Tomatidine (Cat. No. 614350) serves as a suitable negative control.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Induces cyclopia by blocking shh (Sonic Hedgehog) signaling
Product does not compete with ATP.
Reversible: no
Target IC50: ~ 500-700 nM blocking shh (Sonic Hedgehog) signaling in s12 cells

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Preparation Note

Slight warming may be required to achieve complete solubilization in ethanol.

Reconstitution

Following reconstitution in fresh DMSO, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Williams, J.A., et al. 2003. Proc. Natl. Acad. Sci. USA100, 4616.
Cooper, M. K., et al. 2003. Nat. Genet.33, 508.
Mistretta, C.M., 2003. Dev. Biol.254, 1.
ten Berge, D., et al. 2001. Development128, 2929.
Cooper, M.K., et al. 1998. Science280, 1603.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service