Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

260921

Sigma-Aldrich

DNA Methyltransferase Inhibitor II, SGI-1027

Synonym(s):

DNA Methyltransferase Inhibitor II, SGI-1027, N-(4-(2-Amino-6-methylpyrimidin-4-ylamino)phenyl)-4-(quinolin-4-ylamino)-benzamide, S-1027, DNA MTase Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H23N7O
Molecular Weight:
461.52
UNSPSC Code:
12352200

Assay

≥88% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable quinoline compound that is shown to inhibit human DNMT1 (IC50 = 6 and 12.5 µM, respectively, with Hemimethylated DNA or Poly(dI-dC) as the substrate), murine DNMT3a & 3b (IC50 = 8 and 7.5 µM, respectively, with Poly(dI-dC) as the substrate), as well as CpG DNA methylase M.SssI (IC50 = 13.5 and 16 µM, respectively, with Poly(dI-dC) as the substrate and 75 or 150 nM AdoMet) of mycoplasma Spiroplasma sp. strain MQ1, presumably by competing with AdoMet (S-adenosyl-L-methionine , SAM) for the enzyme′s cofactor binding site. Shown to be 2-, 3-, and 5-fold more effective than Decitabine (Cat. No. 189825), respectively, in reactivating p16 mRNA (2.5 µM for 7 days), MLH1 mRNA (2.5 µM for 7 days), and p16 protein expression (1.0 or 2.5 µM for 12 days) in human colon carcinoma RKO cultures by reducing CpG island hypermethylation found in the promoter regions of TSGs (tumor suppressor genes) without inducing genome-wide hypomethylation or caspase-3 activation.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Datta, J., et al. 2009. Cancer Res.69, 4277.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service