Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

240896

Sigma-Aldrich

Cystatin C, Human Urine

A low molecular weight non-glycosylated basic protein of the cystatin superfamily of cysteine protease inhibitors that it is freely filtered by the glomerular membrane.

Synonym(s):

Post γ-globulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

Assay

≥95% (SDS-PAGE)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

0.1 M sodium acetate, pH 4.5: soluble

shipped in

ambient

storage temp.

2-8°C

General description

A low molecular weight non-glycosylated basic protein of the cystatin superfamily of cysteine protease inhibitors that is freely filtered by the glomerular membrane. Serum levels of cystatin C can be used as a specific and sensitive marker of glomerular filtration rate. Protects organisms against undesirable proteolysis by endogenous or exogenous enzymes. Rapidly and tightly inhibits papain (Ki = 0.26 nM) and cathepsin B (Ki = 4.4 nM). Thought to be linked to many disease processes including Alzheimer′s Disease, hereditary cerebral hemorrhage and lung cancer.
A low molecular weight non-glycosylated basic protein of the cystatin superfamily of cysteine protease inhibitors that it is freely filtered by the glomerular membrane. Serum levels of cystatin C can be used as a specific and sensitive marker of glomerular filtration rate. Protects organisms against undesirable proteolysis by endogenous or exogenous enzymes. Thought to be linked to many disease processes including Alzheimer’s disease, hereditary cerebral hemorrhage, and lung cancer.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Cysteine protease
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 20 mM NH₄HCO₃.

Preparation Note

Prepared from the urine of patients with chronic renal tubular proteinuria that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, store in the refrigerator at 4°C. Stock solutions are stable for up to 2 month when stored at -20°C.

Other Notes

Hall, A., et al. 1998. Biochemistry37, 4071.
Peloille, S., et al. 1997. Eur. J. Biochem. 244, 140.
Cimerman, N., et al. 1996. Biol. Chem. Hoppe Seyler 377, 19.
Tenstad, O., et al. 1996. Scand. J. Clin. Lab. Invest.56, 409.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service