Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.93027

Supelco

DL-Lactic acid 88%

for analysis EMPARTA®

Synonym(s):

DL-Lactic acid, 2-Hydroxypropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)COOH
CAS Number:
Molecular Weight:
90.08
Beilstein:
1209341
MDL number:
UNSPSC Code:
12352106

refractive index

n20/D 1.425 (lit.)

bp

122 °C/15 mmHg (lit.)

density

1.209 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

CC(O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)

InChI key

JVTAAEKCZFNVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93027.0521: 500 g, Minimum order quantity: 1
1.93027.2521: 2.5 kg, Minimum order quantity: 1

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service