Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1468180006

Millipore

Tryptic Soy Broth Ready-to-use Bottles

bottle capacity 500 mL, bottle filling volume 450 mL, closure type, blue screw cap with septum and protector, box of 6 bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type blue screw cap with septum and protector
ready-to-use

packaging

box of 6 bottle

technique(s)

direct inoculation: suitable

bottle capacity

500 mL

bottle filling volume

450 mL

pH

7.3 ± 0.2

application(s)

cosmetics
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

General description

Tryptic Soy Broth (TSB) is a complex nutrient rich inhibitor free microbial media designed in compliance with European (EP), Japanese (JP) and United States Pharmacopeia (USP).
The medium is supplemented with peptones from casein and soymeal to provide an enhanced carbon source, which promotes the growth of bacteria, yeast, and fungi present in the test sample.
The medium supports the growth of a wide range of aerobic microbes

Application

Tryptic Soy Broth (TSB) is used for the isolation and cultivation of fastidious bacteria, yeast, and fungi. TSB is suitable for sterility testing of substances, preparations, and products from pharmaceutical and food industry.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service