Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.46640

Millipore

Tryptic Soy Broth Ready-to-use Bottles

Histidine 0.1 %, Cysteine 0.1 %, with Tween® 3 %, Lecithin 0.3 %, bottle capacity 250 mL, bottle filling volume 250 mL, closure type, yellow screw cap with 3 loci, box of 6 bottles

Synonym(s):

CASO, Casein Soybean Digest Broth with 3% Tween, 0.3% Lecithin, 0.1% Histidin, and 0.1% Cysteine, Inactivator Broth, SCDB, TSB, Tryptic Soy Broth + TLHC, Tryptic Soy Broth with neutralizer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type yellow screw cap with 3 loci
ready-to-use

composition

Cysteine, 0.1%
Histidine, 0.1%
Lecithin, 0.3%
with Tween®, 3%

packaging

box of 6 bottles

bottle capacity

250 mL

bottle filling volume

250 mL

pH

7.3±0.2

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Broth with Neutralizers (Tween® 80, Lecithin, Histidine and Cystine) is a universal nutrient rich inhibitor free microbial media designed in compliance with European (EP), Japanese (JP) and United States Pharmacopeia (USP).
The medium is supplemented with peptones from casein and soymeal to provide an enhanced carbon source, which promotes the growth of the microbes present in the test sample.
The neutralizing additives, Polysorbate (Tween®) 80, cysteine, histidine, and lecithin reverse the growth inhibitory effect of most disinfectants and antiseptics. Neutralizing efficiency of histidine is effective against formaldehyde and formaldehyde releasing agents.
Cysteine is a reducing agent and neutralizes oxidizing agents such as hydrogen peroxide and iodine. Lecithin and Polysorbate 80 together are effective against quaternary ammonium compounds, amphoteric surfactants, benzamidines, chlorhexidines and dequadin. Additionally, Polysorbate 80 inactivates benzyl alcohol, carbanilides, dichlorbenzyl alcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols, phenylethyl alcohols and Solbrols (PHB esters).

Application

Tryptic Soy Broth with Neutralizers (Tween® 80, Lecithin, Histidine and Cystine) is used for the isolation and cultivation of fastidious bacteria, yeast. TSB is suitable for sterility testing of substances, preparations, and products from pharmaceutical and food industry.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service