Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

126705-M

Millipore

Anti-Albumin Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

chicken

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

chicken ... Alb(396197)

General description

Ammonium sulfate-purified rabbit polyclonal antibody. Recognizes the ~45 kDa ovalbumin protein.
Anti-Albumin, rabbit polyclonal, reecognizes the ~45 kDa ovalbumin. Exhibits a single arc by immunoelectrophoresis against chicken egg albumin. It is validated for immunoprecipitation.
Recognizes the ~45 kDa ovalbumin protein.
  • Antibody Target Gene Symbol: ALB
  • Target Synonym: ALB1, ALBUMIN, Albumin 1, Albumin isomer 1, Albumin isomer 2, Albumin isomer 3, Albza, DKFZp779N1935, PRO0883, PRO0903, PRO1341, SA, SERUM ALBUMIN, SERUM ALBUMIN CHAIN A, Serum albumin precursor
  • Entrez Gene Name: albumin
  • Hu Entrez ID: 213 (Related Antibodies: 126584, 507313)
  • Mu Entrez ID: 11657
  • Rat Entrez ID: 24186
  • Immunogen

    Chicken Egg
    purified chicken albumin

    Warning

    Toxicity: Standard Handling (A)

    Physical form

    In PBS.

    Reconstitution

    Following initial thaw, aliquot and freeze (-20°C).

    Other Notes

    Exhibits a single arc by immunoelectrophoresis against chicken egg albumin. Variables associated with assay conditions will dictate the proper working dilution.

    Legal Information

    CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

    Not finding the right product?  

    Try our Product Selector Tool.

    Storage Class Code

    12 - Non Combustible Liquids

    WGK

    nwg

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service