Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

06-1352-I

Sigma-Aldrich

Anti-acetyl Histone H4 (Lys12) Antibody

0.5 mg/mL, from rabbit

Synonym(s):

Histone H4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

concentration

0.5 mg/mL

technique(s)

dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys12)

Gene Information

General description

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The histones have an amino terminal tail, a globular domain, and a carboxy-terminal tail. The four core histones, H2A, H2B, H3 and H4 assemble into the octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped in a sequence-independent manner around the octamer, forming the basic subunit of chromatin, the nucleosome. Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail, H4 is involved with the structure of the nucleosomes of the ′beads on a string′ structure.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H4 acetylated at Lys12.

Application

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected acetyl Histone H4 (Lys12) in HeLa cells.

Dot Blot (Specificity) Analysis: 0.1 µg/mL from a representative lot detected acetyl Histone H4 (Lys12) in an Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and in an Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
This Anti-acetyl Histone H4 (Lys12) Antibody is validated for use in western blotting, ICC & dot blot for the detection of acetyl Histone H4 (Lys12).

Quality

Evaluated by Western Blotting in sodium butyrate treated HeLa acid extract.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected acetyl Histone H4 (Lys12) in 10 µg of sodium butyrate treated HeLa acid extract.

Target description

~13 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service