Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V-081

Supelco

(+)-alpha-Tocopheryl-D6 acetate (Vitamin E-D6 acetate) solution

500 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H46D6O3
CAS Number:
Molecular Weight:
478.78
UNSPSC Code:
12352205

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

500 μg/mL in methanol

format

single component solution

storage temp.

−20°C

SMILES string

CC1=C(O[C@@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)CC2)C2=C(C(OC(C)=O)=C1C([2H])([2H])[2H])C([2H])([2H])[2H]

General description

This stable-labeled internal standard is suited for quantitation of tocopheryl acetate by LC/MS or GC/MS Vitamin E testing applications such as quality control testing of Vitamin E fortified foods, dietary supplements, or pharmaceuticals. Tocopheryl acetate is a synthetic form of Vitamin E used as a fortifying agent, dietary supplement for Vitamin E in food or additive in vaping liquids. The U.S. Centers for Disease Control and Prevention (CDC) identified vitamin E acetate as very strong culprit of concern in the recent vaping related lung disease outbreak in the United States.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service