Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z611662

Julabo® bath accessories

Aqua-Stabil, water bath protective media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103706
NACRES:
NB.77

packaging

pk of 6 × 100 mL

manufacturer/tradename

Julabo 8940006

availability

not available in USA and Canada

Looking for similar products? Visit Product Comparison Guide

Application

Add 2 mL of Aqua-Stabil to a liter of water. Whenever the blue tinting disappears (or no later than three months), fill the bath with fresh water and Aqua-Stabil.

Legal Information

Julabo is a registered trademark of Julabo USA, Inc.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service