Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z308986

Quickfit® ground sleeve stirrer glands

shaft D 11 mm, joint: ST/NS 29/32

Synonym(s):

ST20/4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.72

Joint

joint: ST/NS29/32

manufacturer/tradename

Quickfit ST 20/4

availability

not available in USA and Canada

shaft D

11 mm

fitting

24 thread

Looking for similar products? Visit Product Comparison Guide

General description

Shaft that passes through screw thread adapterr in top of gland. Will operate at up to 700rpm and down to 0.1 torr. Cup shaped lubricant reservoir surrounds sleeve such that lubricant also acts as coolant.

Legal Information

Quickfit is a registered trademark of SciLabware Limited

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service