Key Documents
TEM-CF200AU50
Sigma-Aldrich
Carbon Film Supported Gold Square Mesh
size 200 mesh, box of 50
Synonym(s):
CRBON FILM 200 MESH AU 50/BX
About This Item
UNSPSC Code:
41111761
packaging
box of 50
size
200 mesh
General description
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Gold grid, 200 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)