Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QCRBNSET

Sigma-Aldrich

QuicTPD CRBN ligand Set

new
Sign Into View Organizational & Contract Pricing


About This Item

packaging

pkg of 50 mg

Quality Level

storage temp.

2-8°C

General description

5-Amino Thalidomide, Lenalidomide, Pomalidomide and C-5 Lenalidomide are recruiters of CRBN protein that can be leveraged for targeted protein degradation research. They are provided at 50 mg each.

Application

Useful for construction of CRBN based partial protacs of full protacs, by utilizing the amine terminus to add suitable linkers with peptide coupling reactions. These can also be used as controls/competitors to confirm target engagement of putative protacs.

Features and Benefits

Quantities provided are sufficient to do atleast 96 reactions at 2 micromole each with a D2B approach with a library of suitable bifunctional linkers ending in carboxylic acid terminus to generate a library of partial protacs or full protacs.

Other Notes

CRBN Ligand Set part of QuicTPD kit

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Repr. 1A - STOT RE 2

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service