Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF86468754

Tantalum

foil, 0.1m coil, thickness 0.03mm, coil width 1.2mm, annealed, 99.9%

Synonym(s):

Tantalum, TA000354, Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 537.2 °C)

Assay

≥99.9%

form

foil

autoignition temp.

572 °F

manufacturer/tradename

Goodfellow 864-687-54

resistivity

13.5 μΩ-cm, 20°C

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingxun Du et al.
Journal of hazardous materials, 225-226, 21-27 (2012-05-23)
In order to overcome the limitation of the application of nanoparticles, tantalum (oxy)nitrides nanotube arrays on a Ta foil were synthesized and introduced in vis (visible light)-Fenton-like system to enhance the degradation of atrazine. At first, the anodization of tantalum
Zhongli Cai et al.
The journal of physical chemistry. B, 109(10), 4796-4800 (2006-07-21)
Both monolayer and thick (20 microm) films of dry pGEM-3Zf(-) plasmid DNA deposited on tantalum foil were exposed to Al Kalpha X-rays (1.5 keV) for various times in an ultrahigh vacuum chamber. For monolayer DNA, the damage was induced mainly
J G Sen Gupta
Talanta, 31(12), 1053-1056 (1984-12-01)
A 40-fold increase in sensitivity obtained by using a tantalum foil lining in a pyrolytically-coated graphite furnace permitted determination of low ppm levels of cerium in most silicate rocks. A preliminary preconcentration by oxalate and hydroxide co-precipitations was used before
Zhongli Cai et al.
Radiation research, 165(3), 365-371 (2006-02-24)
Both thick and thin films of pGEMR-3Zf- plasmid DNA deposited on a tantalum foil were exposed to soft X rays (effective energy of 14.8 keV) for various times in air under a relative humidity of 45% (Gamma approximately 6, where
J G Sen Gupta
Talanta, 32(1), 1-6 (1985-01-01)
An improved graphite furnace atomic-absorption method has been developed for the determination of Sc, Y and the rare-earth elements in silicate rocks and related materials. The method, which involves the separation of the lanthanides by ion-exchange followed by their determination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service