Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF63236213

Cadmium

foil, 15mm disks, thickness 0.05mm, as rolled, 99.99+%

Synonym(s):

Cadmium, CD000200

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cd
CAS Number:
Molecular Weight:
112.41
Beilstein:
8137359
MDL number:
UNSPSC Code:
12141707
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 632-362-13

resistivity

7.27 μΩ-cm, 22°C

bp

765 °C (lit.)

mp

320.9 °C (lit.)

density

8.65 g/mL at 25 °C (lit.)

SMILES string

[Cd]

InChI

1S/Cd

InChI key

BDOSMKKIYDKNTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen F Kry et al.
Journal of applied clinical medical physics, 8(4), 2679-2679 (2008-05-02)
The commonly used thermoluminescent dosimeter TLD-100 (Harshaw Chemical Company, Solon, OH) responds not only to photons and electrons, but also to neutrons that are produced during high-energy therapies. As a result, TLD-100 measurements outside of the treatment field are suspect
Y Nakamura et al.
Radioisotopes, 28(11), 675-680 (1979-11-01)
Formation cross sections and thick-target yields were studied in detail for alpha reactions and helium-3 reactions of natural cadmium in the energy range 10-40 MeV in order to determine the optimum condition for the production of medical use 11In. The
Uroš Cvelbar et al.
Nanotechnology, 19(40), 405605-405605 (2008-10-08)
Plasma-assisted synthesis of nanostructures is one of the most precise and effective approaches used in nanodevice fabrication. Here we report on the innovative approach of synthesizing nanostructured cadmium oxide films on Cd substrates using a reactive oxygen plasma-based process. Under
Jian-An Xian et al.
Bulletin of environmental contamination and toxicology, 92(5), 525-528 (2014-01-01)
This study investigated the effect of ambient Cadmium (Cd) on haemocyte apoptosis of the shrimp, Penaeus monodon. Cellular response was determined in Cd-exposed (0, 0.05, 0.5 and 5 mg L(-1)) shrimp. Results showed that 0.05 mg L(-1) Cd(2+) had no significant effect on the
Peerapat Kosolsaksakul et al.
Environmental pollution (Barking, Essex : 1987), 187, 153-161 (2014-02-08)
The Mae Tao watershed, northwest Thailand, has become contaminated with cadmium (Cd) as a result of zinc ore extraction (Padaeng deposit) in the nearby Thanon-Thongchai mountains. Consumption of contaminated rice has led to documented human health impacts. The aim of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service