Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF50394269

Inconel® alloy 600 - heat resisting alloy, Ni72Cr16Fe8

1000mm tube, 9.5mm outside diameter, 1.7mm wall thickness, 6.1mm inside diameter, annealed

Synonym(s):

NI027450

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 503-942-69

L × wall thickness

1000 mm × 1.7 mm

O.D. × I.D.

9.5 mm × 6.1 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service