Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF43252848

Invar® - Controlled Expansion Alloy, Fe64Ni36

foil, 25 x 25mm, 0.125mm thickness, half hard

Synonym(s):

FE020265

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

form

foil (temper half hard)

manufacturer/tradename

Goodfellow 432-528-48

size × thickness

25 mm × 25 mm × 0.125 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Invar is a registered trademark of Aperam Alloys Imphy
Invar is a registered trademark of Aperam Alloys Imphy

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service