GF38827682
Carbon
microleaf, 25x70mm, thinness 0.025μm, specific density 5μg/cm2, temporary glass support, 99.997%
Synonym(s):
Activated charcoal, C 004910, Charcoal activated
About This Item
vapor pressure
<0.1 mmHg ( 20 °C)
Assay
99.997%
form
foil (microleaf)
autoignition temp.
842 °F
manufacturer/tradename
Goodfellow 388-276-82
resistivity
1375 μΩ-cm, 20°C (graphite)
size × thickness
25 × 70 mm × 0.025 μm
mp
3550 °C (lit.)
SMILES string
[C]
InChI
1S/C
InChI key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service