Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF27731853

Nickel

wire reel, 10m, diameter 0.05mm, as drawn, 99%

Synonym(s):

Nickel, NI005110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

wire

manufacturer/tradename

Goodfellow 277-318-53

resistivity

6.97 μΩ-cm, 20°C

L × diam.

10 m × 0.05 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Egor Geist et al.
Natural product reports, 31(4), 441-448 (2014-02-28)
This Highlight covers the current status of relatively unexplored sp(3)-sp(3) cross-coupling reactions with particular focus on natural product and related syntheses.
Gonensin O Bozdag et al.
Gene, 538(2), 251-257 (2014-02-04)
Nickel is an essential micronutrient due to its involvement in many enzymatic reactions as a cofactor. However, excess of this element is toxic to biological systems. Here, we constructed a cDNA library from Beta maritima and screened it in the
Giovanna Visioli et al.
Bulletin of environmental contamination and toxicology, 92(4), 490-496 (2013-11-30)
In vitro short-term chronic phytotoxicity germination and root elongation test were applied to test the effects of nickel (Ni) in seed germination and root elongation in six plants species: Cucumis sativus (Cucurbitaceae), Lepidium sativum and Brassica nigra (Brassicaceae), Trifolium alexandrinum
Daniel H Rothman et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(15), 5462-5467 (2014-04-08)
The end-Permian extinction is associated with a mysterious disruption to Earth's carbon cycle. Here we identify causal mechanisms via three observations. First, we show that geochemical signals indicate superexponential growth of the marine inorganic carbon reservoir, coincident with the extinction
Gary A Molander et al.
Organic letters, 16(7), 1904-1907 (2014-03-29)
A general method for the alkenylation of alkyl electrophiles using nearly stoichiometric amounts of the air- and moisture-stable potassium organotrifluoroborates has been developed. Various functional groups were tolerated on both the nucleophilic and electrophilic partner. Reactions of highly substituted E-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service