Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900304

Sigma-Aldrich

CdS/ZnS core-shell type quantum dots

amine functionalized, fluorescence λem 450 nm, 4 μM in PBS

Synonym(s):

Fluorescent nanocrystals, QDs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

form

dispersion

concentration

4 μM in PBS

λmax

435 nm

fluorescence

λem 450 nm
λem 450 nm±10 nm, quantum yield <50%

functional group

amine

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

The amine functionalized CdS/ZnS or CdSe/ZnS quantum dots (QDs) find applications in bioimaging and biosensing. The three main conjugation approaches reported in literature are conjugation using glutaraldehyde as a linker, conjugations with carboxylic acid containing molecules by using 1-ethyl-3-(3-dimethylaminopropyl) carbodiimide (EDC) coupling, and direct conjugation with isothiocyanates bearing molecules.

Caution

Do not freeze.

Legal Information

Product of Ocean Nanotech, LLC.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kumar P, et al.
Sensors and Actuators B, Chemical, 202, 404-404 (2014)
Jian Wei Tay et al.
Scientific reports, 4, 3918-3918 (2014-01-30)
Phase distortions due to scattering in random media restrict optical focusing beyond one transport mean free path. However, scattering can be compensated for by applying a correction to the illumination wavefront using spatial light modulators. One method of obtaining the
Lily Yang et al.
Small (Weinheim an der Bergstrasse, Germany), 5(2), 235-243 (2008-12-18)
Epidermal growth factor receptor (EGFR) targeted nanoparticle are developed by conjugating a single-chain anti-EGFR antibody (ScFvEGFR) to surface functionalized quantum dots (QDs) or magnetic iron oxide (IO) nanoparticles. The results show that ScFvEGFR can be successfully conjugated to the nanoparticles
Kavitha Pathakoti et al.
Journal of environmental sciences (China), 25(1), 163-171 (2013-04-17)
Quantum dots (QD) nanoparticles have been widely used in biomedical and electronics fields, because of their novel optical properties. Consequently it confers enormous potential for human exposure and environmental release. To increase the biocompatibility of QDs, a variety of surface

Articles

Professor Xiaohu Gao (University of Washington, USA) provides a overview of recent quantum dot (QD) advancements and their potential for advancing bioassay and bioimaging technologies.

Professor Sharma and colleagues review the synthesis and applications of this novel material. This includes a discussion of the unique properties of quantum dots and their suitability for solar cell applications, along with common synthesis techniques used to develop these materials.

Synthesis of Halide Perovskite Quantum Dots for Display Applications

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service