Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808229

Sigma-Aldrich

Aluminum-doped zinc oxide nanoparticle ink

2.5 wt. %, viscosity 40 cP, work function -3.9eV

Synonym(s):

AZO ink, Al-doped ZnO dispersion, Al-dopend ZnO suspension, Al:ZnO ink, Avantama N-21X-Screen, Nanograde N-21X-Screen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103

form

dispersion

concentration

2.5 wt. % (crystalline Al doped ZnO (3.15 mol% Al) in mixture of alcohols)

work function

-3.7--4.1 eV

color

translucent gray-brown

particle size

8-16 nm

viscosity

32-48 cP

Looking for similar products? Visit Product Comparison Guide

Application

Al-doped ZnO nanoparticle ink is for screen printing for the use as electron transport layer in printed electronics. Al-doped ZnO nanoparticle ink is universally applicable in normal and inverted architecture with performance comparable to evaporated calcium.

Preparation Note

  • Storage: In dark at room temperature
  • Prior to application: Shake, ultrasonicate with sonic horn and (optionally) filter through 0.45μm PTFE filter.
  • Post-treatment: Annealing of deposited Al:ZnO films at >100°C.

Legal Information

Product of Avantama Ltd.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

120.2 °F

Flash Point(C)

49 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professor Tokito and Professor Takeda share their new materials, device architecture design principles, and performance optimization protocols for printed and solution-processed, low-cost, highly flexible, organic electronic devices.

Recent progress in the area of solution-processed functional materials has led to the development of a variety of thin-film optoelectronic devices with significant promise in the industrial and consumer electronics fields.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service