Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

750069

Sigma-Aldrich

Poly(ethylene glycol) methyl ether 2-bromoisobutyrate

average Mn 2,000

Synonym(s):

ATRP initiator, PEG methyl ether bromoisobutyrate, mPEG bromoisobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)nCOC(CH3)2Br
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

mol wt

average Mn 2,000

transition temp

Tm 47-52

Mw/Mn

≤1.2

Looking for similar products? Visit Product Comparison Guide

Application

Poly(ethylene glycol)-containing ATRP initiator for generating a hydrophilic block, as well as enhancing biocompatibility.

Other Notes

average PEG Mn 2,000; n ~44

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jakubowski, W.; Tsarevsky, N. V.; McCarthy, P.; Matyjaszewski, K.
Material Matters, 5, 16-16 (2010)

Articles

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

Applying ARGET ATRP to the Growth of Polymer Brush Thin Films by Surface-initiated Polymerization

We presents an article about Copper(I)-mediated Living Radical Polymerization in the Presence of Pyridylmethanimine Ligands, and the emergence of living radical polymerization mediated by transition metal catalysts in 1995, which was a seminal piece of work in the field of synthetic polymer chemistry.

Protocols

Sigma-Aldrich presents an article about the typical procedures for polymerizing via ATRP, which demonstrates that in the following two procedures describe two ATRP polymerization reactions as performed by Prof. Dave Hadddleton′s research group at the University of Warwick.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service