Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

704237

Sigma-Aldrich

Lanthanum strontium manganite

greener alternative

composite cathode powder, LSM-20/GDC10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NA.23

form

solid

composition

Ce0.9Gd0.1O2, 50 wt. %
La0.8Sr0.2MnO3, 50 wt. %

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

4-8 m2/g

particle size

0.4-0.9 μm (Horiba-centrifugal)

application(s)

battery manufacturing

greener alternative category

Looking for similar products? Visit Product Comparison Guide

Application

LSM is one of the materials that is useful as an electrode material in solid oxide fuel cells.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of lanthanum strontium manganite perovskite cathode materials of solid oxide fuel cells: a review.
Jiang SP
J. Mater. Sci., 43(21), 6799-6833 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service