Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

689629

Sigma-Aldrich

2-Amino-4-bromophenol

≥98.0% (HPLC)

Synonym(s):

4-Bromo-2-aminophenol, NSC 523846

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6BrNO
CAS Number:
Molecular Weight:
188.02
Beilstein:
3236448
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

≥98.0% (HPLC)

form

lumps

mp

130-135 °C

SMILES string

Nc1cc(Br)ccc1O

InChI

1S/C6H6BrNO/c7-4-1-2-6(9)5(8)3-4/h1-3,9H,8H2

InChI key

JHRIPENGTGSNPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of (aryl)oxadiazolobenzoxazinones via Suzuki-Miyaura reaction
  • Copper-catalyzed oxidative amination of benzoxazoles and related azoles via C-H and C-N bond activation
  • Synthesis of N-(alkoxyphenyl)-aminocarbonylbenzoic acid derivatives as protein tyrosine phophatase 1B inhibitors
  • Preparation of lipophilic deoxy-xylulose-phosphate reductoisomerase inhibitors
  • Synthesis of benzoxazole benzenesulfonamides as allosteric inhibitors of fructose-1,6-bisphosphatase

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service