Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

671533

Sigma-Aldrich

(1R,2R)-1-Amino-2-benzyloxycyclopentane

97%

Synonym(s):

(1R)-trans-2-(Phenylmethoxy)cyclopentanamine, (1R,2R)-trans-2-Benzyloxy-cyclopentylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO
CAS Number:
Molecular Weight:
191.27
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.5% (GC)
97%

optical purity

enantiomeric excess: ≥98.0%

storage temp.

2-8°C

SMILES string

N[C@@H]1CCC[C@H]1OCc2ccccc2

InChI

1S/C12H17NO/c13-11-7-4-8-12(11)14-9-10-5-2-1-3-6-10/h1-3,5-6,11-12H,4,7-9,13H2/t11-,12-/m1/s1

InChI key

JIMSXLUBRRQALI-VXGBXAGGSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service