Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

661007

Sigma-Aldrich

Hydroxy[-(S)-BINAP]-rhodium(I) Dimer

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C88H66O2P4Rh2
Molecular Weight:
1485.17
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

reaction suitability

reaction type: click chemistry

impurities

3-5% THF
3-5% ethanol

SMILES string

[H]O[Rh].[H]O[Rh].c1ccc(cc1)P(c2ccccc2)c3ccc4ccccc4c3-c5c(ccc6ccccc56)P(c7ccccc7)c8ccccc8.c9ccc(cc9)P(c%10ccccc%10)c%11ccc%12ccccc%12c%11-c%13c(ccc%14ccccc%13%14)P(c%15ccccc%15)c%16ccccc%16

InChI

1S/2C44H32P2.2H2O.2Rh/c2*1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38;;;;/h2*1-32H;2*1H2;;/q;;;;2*+1/p-2

InChI key

PWIUOLBVYNHREE-UHFFFAOYSA-L

Application

Catalyst employed in an asymmetric 1,6-addition of arylzinc reagents to dienones leading to chiral 5-substituted enones. Also used in an asymmetric 1,4-addition of organoboron reagents to cyclic and acyclic α,β-unsaturated ketones with excellent enantioselectivity.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhodium-catalyzed asymmetric 1,6-addition of aryl zinc reagents to dienones.
Tamio Hayashi et al.
Angewandte Chemie (International ed. in English), 44(27), 4224-4227 (2005-06-02)
Rhodium-catalyzed asymmetric 1,4-addition and its related asymmetric reactions.
Tamio Hayashi et al.
Chemical reviews, 103(8), 2829-2844 (2003-08-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service