Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

655546

Sigma-Aldrich

1,6-Bis(p-carboxyphenoxy)hexane

90%

Synonym(s):

CPH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CC6H4O(CH2)6OC6H4CO2H
CAS Number:
Molecular Weight:
358.39
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

mp

278-290 °C

functional group

carboxylic acid

SMILES string

OC(=O)c1ccc(OCCCCCCOc2ccc(cc2)C(O)=O)cc1

InChI

1S/C20H22O6/c21-19(22)15-5-9-17(10-6-15)25-13-3-1-2-4-14-26-18-11-7-16(8-12-18)20(23)24/h5-12H,1-4,13-14H2,(H,21,22)(H,23,24)

InChI key

FQJXYULOQZUKBZ-UHFFFAOYSA-N

Application

Building block for polyanhydrides, a class of bioerodible polymer with drug delivery applications.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catriona P Harkins et al.
Genome biology, 18(1), 130-130 (2017-07-21)
The spread of drug-resistant bacterial pathogens poses a major threat to global health. It is widely recognised that the widespread use of antibiotics has generated selective pressures that have driven the emergence of resistant strains. Methicillin-resistant Staphylococcus aureus (MRSA) was
Zeb R Zacharias et al.
Frontiers in immunology, 9, 1953-1953 (2018-09-21)
Influenza A virus (IAV) is a major cause of respiratory illness. Given the disease severity, associated economic costs, and recent appearance of novel IAV strains, there is a renewed interest in developing novel and efficacious "universal" IAV vaccination strategies. Recent
Yu Chong et al.
Environmental science & technology, 51(17), 10154-10161 (2017-08-05)
Before graphene derivatives can be exploited as next-generation antimicrobials, we must understand their behavior under environmental conditions. Here, we demonstrate how exposure to simulated sunlight significantly enhances the antibacterial activity of graphene oxide (GO) and reveal the underlying mechanism. Our
Yan-Lin He et al.
PloS one, 7(7), e41303-e41303 (2012-07-31)
Cyproheptadine (CPH) is a histamine- and serotonin-receptor antagonist, and its effects are observed recently in the modulation of multiple intracellular signals. In this study, we used cortical neurons and HEK-293 cells transfected with Kv2.1 α-subunit to address whether CPH modify
Binbin Shao et al.
Journal of hazardous materials, 362, 318-326 (2018-09-23)
In this study, the hollow mesoporous carbon spheres (HMCs) were synthesized and modified for laccase (Lac) immobilization, and the structural characteristics of HMCs materials were determined by FESEM, TEM and FTIR etc. The maximum loading of Lac on the HMCs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service