Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

642355

Sigma-Aldrich

2-Chloro-5-nitrobenzanilide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)CONHC6H5
CAS Number:
Molecular Weight:
276.68
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

158-159 °C (lit.)

functional group

amide
chloro
nitro

SMILES string

[O-][N+](=O)c1ccc(Cl)c(c1)C(=O)Nc2ccccc2

InChI

1S/C13H9ClN2O3/c14-12-7-6-10(16(18)19)8-11(12)13(17)15-9-4-2-1-3-5-9/h1-8H,(H,15,17)

InChI key

DNTSIBUQMRRYIU-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Giampietri et al.
Biochimica et biophysica acta, 1851(7), 929-936 (2015-03-10)
c-Flip proteins are well-known apoptosis modulators. They generally contribute to tissue homeostasis maintenance by inhibiting death-receptor-mediated cell death. In the present manuscript, we show that c-Flip knock-out (KO) mouse embryonic fibroblasts (MEFs) kept in culture under starvation conditions gradually modify
Y Xu et al.
Oncogene, 34(32), 4229-4237 (2014-11-05)
Manganese superoxide dismutase (MnSOD) is a mitochondrially localized primary antioxidant enzyme, known to be essential for the survival of aerobic life and to have important roles in tumorigenesis. Here, we show that MnSOD deficiency in skin tissues of MnSOD-heterozygous knockout
Meng Xu et al.
Cell, 178(6), 1478-1492 (2019-09-03)
Liver fibrosis is a very common condition seen in millions of patients with various liver diseases, and yet no effective treatments are available owing to poorly characterized molecular pathogenesis. Here, we show that leukocyte cell-derived chemotaxin 2 (LECT2) is a
Masahiro Myojo et al.
PloS one, 9(5), e96948-e96948 (2014-05-16)
Because endothelial nitric oxide synthase (eNOS) has anti-inflammatory and anti-arteriosclerotic functions, it has been recognized as one of the key molecules essential for the homeostatic control of blood vessels other than relaxation of vascular tone. Here, we examined whether telmisartan
Xuxia Sui et al.
PloS one, 9(10), e110828-e110828 (2014-10-22)
EP3, one of four prostaglandin E2 (PGE2) receptors, is significantly lower in atherosclerotic plaques than in normal arteries and is localized predominantly in macrophages of the plaque shoulder region. However, mechanisms behind this EP3 expression pattern are still unknown. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service