Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

631884

Sigma-Aldrich

Barium zirconate

nanopowder, <50 nm particle size, 98.5% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaZrO3
CAS Number:
Molecular Weight:
276.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

98.5% trace metals basis

form

nanopowder

reaction suitability

reagent type: catalyst
core: zirconium

particle size

<50 nm

density

5.52 g/mL at 25 °C (lit.)

SMILES string

[Ba++].[O-][Zr]([O-])=O

InChI

1S/Ba.3O.Zr/q+2;;2*-1;

InChI key

DQBAOWPVHRWLJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of nanosized barium zirconate powder by thermal decomposition of urea in an aqueous solution containing barium and zirconium, and by calcination of the precipitate.
Boschini F,et al.
J. Eur. Ceram. Soc., 23(16), 3035-3042 (2003)
Synthesis and sintering of large batches of barium zirconate nanopowders.
Sin A, et al.
Journal of the American Ceramic Society. American Ceramic Society, 85(8), 1928-1932 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service