Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

572691

Sigma-Aldrich

3,4′-5-Trihydroxystilbene-3-β-D-glucopyranoside

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22O8
CAS Number:
Molecular Weight:
390.38
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D −55°, c = 1 in acetone

mp

223-226 °C (lit.)

SMILES string

OC(C=C1)=CC=C1/C=C/C2=CC(O[C@H]3[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O3)=CC(O)=C2

InChI

1S/C20H22O8/c21-10-16-17(24)18(25)19(26)20(28-16)27-15-8-12(7-14(23)9-15)2-1-11-3-5-13(22)6-4-11/h1-9,16-26H,10H2/b2-1+/t16-,17-,18+,19-,20-/m1/s1

InChI key

HSTZMXCBWJGKHG-CUYWLFDKSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Wen et al.
Biomedical microdevices, 14(4), 721-728 (2012-04-25)
Caenorhabditis elegans (C. elegans) is an excellent model organism for the study of aging and longevity. In this work, we presented a microfluidic approach for the evaluation of longevity in C. elegans under stress. The microfluidic device integrated multiple microvalves
M J Wu et al.
International journal of immunopathology and pharmacology, 25(4), 923-934 (2013-01-10)
Fulminant hepatic failure (FHF) is a devastating clinical syndrome with extremely poor prognosis and high mortality. Therefore, better treatment is urgently needed. Polydatin (PD), a traditional anti-inflammatory drug, has been described to protect against liver injury induced by certain hepatotoxins.
Polydatin suppresses ultraviolet B-induced cyclooxygenase-2 expression in vitro and in vivo via reduced production of reactive oxygen species.
Y-D He et al.
The British journal of dermatology, 167(4), 941-944 (2012-03-23)
Maria Rotches-Ribalta et al.
Pharmacological research, 66(5), 375-382 (2012-08-22)
A pharmacokinetic study of the metabolic profile of resveratrol has been performed in healthy men after moderate red wine (RW) consumption. The bioavailability of resveratrol is highly influenced by several factors such as the food matrix and, therefore, this study
Long-Tao Liu et al.
Chinese journal of integrative medicine, 18(9), 714-719 (2012-09-01)
Rhizoma Polygoni Cuspidati, a Chinese herbal drug, has actions of dispelling dampness, alleviating jaundice, clearing heat, subsiding toxin, activating blood, and removing stasis. Polydatin (PD), one of its chief active ingredients, has been proved by modern pharmacological studies to possess

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service