Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

465895

Sigma-Aldrich

Calcium

dendritic pieces, purified by distillation, 99.5% trace metals basis

Synonym(s):

Atomic calcium, Calcium atom, Calcium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ca
CAS Number:
Molecular Weight:
40.08
Beilstein:
4241647
EC Number:
MDL number:
UNSPSC Code:
12141503
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.5% trace metals basis

form

dendritic pieces

purified by

distillation

reaction suitability

reagent type: reductant

resistivity

3.5 μΩ-cm, 20°C

bp

1484 °C (lit.)

mp

850 °C (lit.)

density

1.54 g/mL at 25 °C (lit.)

SMILES string

[Ca]

InChI

1S/Ca

InChI key

OYPRJOBELJOOCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to Ca8Al3 and Ca13Al14; intermetallics with new metallic Zintl phases.7
Solid state synthesis with Mg and Sn resulted in a new phase, Ca6.2Mg3.8Sn7, which has an unprecedented type of tin chain composed of square-planar tin units.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ganguli, A.K. et al.
Journal of the American Chemical Society, 120, 1223-1223 (1998)
Emmanuel Bourinet et al.
Physiological reviews, 94(1), 81-140 (2014-01-03)
The detection and processing of painful stimuli in afferent sensory neurons is critically dependent on a wide range of different types of voltage- and ligand-gated ion channels, including sodium, calcium, and TRP channels, to name a few. The functions of
Tsai-Wen Chen et al.
Nature, 499(7458), 295-300 (2013-07-23)
Fluorescent calcium sensors are widely used to image neural activity. Using structure-based mutagenesis and neuron-based screening, we developed a family of ultrasensitive protein calcium sensors (GCaMP6) that outperformed other sensors in cultured neurons and in zebrafish, flies and mice in
Byrappa Venkatesh et al.
Nature, 505(7482), 174-179 (2014-01-10)
The emergence of jawed vertebrates (gnathostomes) from jawless vertebrates was accompanied by major morphological and physiological innovations, such as hinged jaws, paired fins and immunoglobulin-based adaptive immunity. Gnathostomes subsequently diverged into two groups, the cartilaginous fishes and the bony vertebrates.
S Boonen et al.
Journal of internal medicine, 259(6), 539-552 (2006-05-18)
Combined calcium and vitamin D supplementation is an essential component of the management of osteoporosis, supported by a strong scientific rationale. The types of individuals who should receive calcium and vitamin D supplements are those: (i) patients with documented osteoporosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service