Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

456632

Sigma-Aldrich

Polyethylene-graft-maleic anhydride

viscosity 1,700-4,500 cP (140 °C)(lit.)

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

viscosity

1,700-4,500 cP(140 °C)(lit.)

saponification value

32‑36 mg KOH/g

transition temp

Tm (DSC) 105 °C (at peak)

solubility

H2O: insoluble
toluene and xylene: soluble

InChI

1S/C4H2O3.C2H4/c5-3-1-2-4(6)7-3;1-2/h1-2H;1-2H2

InChI key

YYXLGGIKSIZHSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bond aid for polar to nonpolar substances. Compatibilizer for polymer blends.

Features and Benefits

Contributes abrasion resistance, lubricity, antiblocking, and surface glass, allows chemical bonding via anhydride group.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Baloğlu et al.
Farmaco (Societa chimica italiana : 1989), 58(5), 391-396 (2003-05-06)
Bioadhesive tablet formulations have been developed for mucosal application. Sixteen different bioadhesive tablet formulations were prepared and evaluated. Their bioadhesion to vaginal mucosa were studied by tensile testing method. The swelling behaviour of the tablets in three different solutions was
Sijing Xiong et al.
Archives of toxicology, 87(1), 99-109 (2012-08-14)
To uncover the size influence of TiO(2) nanoparticles on their potential toxicity, the cytotoxicity of different-sized TiO(2) nanoparticles with and without photoactivation was tested. It was demonstrated that without photoactivation, TiO(2) nanoparticles were inert up to 100 μg/ml. On the contrary
H Neumann et al.
Applied biochemistry and biotechnology, 11(3), 177-189 (1985-06-01)
Alkaline phosphatase, APase, (EC 3.1.31) from calf intestine, after shifting the equilibrium by effector molecules towards the dimeric form of the enzyme, was coupled (ratio 1:2, protein: copolymer) to a copolymer of ethylene and maleic anhydride, EMA. The water-soluble APase-EMA
C Minard-Basquin et al.
Nucleosides, nucleotides & nucleic acids, 20(4-7), 895-899 (2001-09-21)
(Polymer-oligonucleotide) conjugates were obtained via direct ON synthesis from the poly(ethylene-alt-maleic anhydride: PEMA) grafted onto glass bead surfaces. The effect of the ON number per polymer chain on binding was evaluated through Tm experiments. Results were correlated with ELOSA (Enzyme
J K Song et al.
Journal of microencapsulation, 24(1), 11-19 (2007-04-19)
This paper shows two types of microcapsules used for electrophoretic display. One is prepared by in-situ polymerization which is based on urea, melamine and formaldehyde and another by complex coacervation, which is composed of gelatin and gum Arabic. Microcapsules attract

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service