Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

455954

Sigma-Aldrich

Chromium(III) sulfate hydrate

99.999% trace metals basis

Synonym(s):

Chromic sulfate hydrate, Chromium trisulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cr2(SO4)3 · xH2O
CAS Number:
Molecular Weight:
392.18 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

solid

impurities

≤20.0 ppm Trace Metal Analysis

mp

100 °C (dec.) (lit.)

solubility

H2O: soluble(lit.)

density

1.7 g/mL at 25 °C (lit.)

SMILES string

O.[Cr+3].[Cr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Cr.3H2O4S.H2O/c;;3*1-5(2,3)4;/h;;3*(H2,1,2,3,4);1H2/q2*+3;;;;/p-6

InChI key

KVIYQADEXNCDCB-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service