Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

432288

Sigma-Aldrich

Candelilla wax

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

beads

bp

>240 °C (lit.)

mp

68-72 °C (lit.)

acid number

11‑19 mg KOH/g

ester number

40‑47

iodine value

19‑44

saponification value

44 mg KOH/g

solubility

ethanol, benzene and petroleum ether (hot): soluble

density

0.988 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

General description

Candelilla Wax is a common wax derived from the leaves of the candelilla shrub. It has a composition which consists of n-alkanes, high molecular weight esters and alcohols, and sterols and free acids.
Candelilla wax is an additive which can be used in cosmetic and food industry.

Application

Component in leather dressings, polishes, varnishes, sealing wax, electrical insulating compositions, paper sizing lubricants and adhesives.

Features and Benefits

Hard glossy wax that retains oil, is emulsifiable and has excellent gelling properties. Low coefficient of expansion, very durable wax, will not crack.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Edible coatings from candelilla wax microemulsions.
HAGENMAIER RD and BAKER RA
Journal of Food Science, 61(3), 562-565 (1996)
Wax-Impregnated Cotton Fabrics as Cell Culture Platform.
Advanced Materials Research, 1112, 441-444 (2015)
Thermal and textural properties of organogels developed by candelilla wax in safflower oil.
Toro-Vazquez JF, et al.
Journal of the American Oil Chemists' Society, 84(11), 989-1000 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service