Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

35248

Sigma-Aldrich

2,3-Dichlorobenzaldehyde

≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CHO
CAS Number:
Molecular Weight:
175.01
Beilstein:
508198
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

mp

64-67 °C (lit.)

SMILES string

[H]C(=O)c1cccc(Cl)c1Cl

InChI

1S/C7H4Cl2O/c8-6-3-1-2-5(4-10)7(6)9/h1-4H

InChI key

LLMLNAVBOAMOEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,3-Dichlorobenzaldehyde is suitable for use in the preparation of:
  • 4-(2,3-dichlorobenzylideneamino)-1,5-dimethyl-2-phenyl-1H-pyrazol-3(2H)-one, Schiff base
  • ethyl 4-(2,3-dichlorophenyl)-1,6-dimethyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate via Biginelli condensation
  • benzyl 4-(2,3-dichlorophenyl)-1,3,6-trimethyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

275.0 °F - closed cup

Flash Point(C)

135 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

4-(2, 3-Dichlorobenzylideneamino)-1, 5-dimethyl-2-phenyl-1H-pyrazol-3 (2H)-one.
Sun, Y-X., et al.
Acta Crystallographica Section E, Structure Reports Online, 62(10), o4613-o4615 (2006)
Synthesis and reactions of Biginelli-compounds. Part 23.1 Chemoenzymatic syntheses of enantiomerically pure 4-aryl-3, 4-dihydropyrimidin-2 (1H)-ones.
Schnell B, et al.
Journal of the Chemical Society. Perkin Transactions 1, 24, 4382-4389 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service